3,5-Dichlorobenzoic acid

Leverancier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

BTB03142.1 BTB03142EA
MAYBBTB03142.1EA 0 EUR
MAYBBTB03142.1 MAYBBTB03142EA
3,5-Dichlorobenzoic acid
3,5-Dichlorobenzoic acid
Formule: C₇H₄Cl₂O₂
Kookpunt: 309 °C (1013 hPa)
Smeltpunt: 183…185 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00002494
CAS nummer: 51-36-5
EINECS: 200-092-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR