2-Chloro-4-fluoroaniline

Leverancier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

BTB05781.1 BTB05781.50
MAYBBTB05781.1EA 0 EUR
MAYBBTB05781.1 MAYBBTB05781.50
2-Chloro-4-fluoroaniline
2-Chloro-4-fluoroaniline
Formule: C₆H₅ClFN
MW: 145.56 g/mol
Kookpunt: 192
Dichtheid: 1.21 g/cm³ (20 °C)
MDL Nummer: MFCD00042530
CAS nummer: 2106-02-7
EINECS: 218-282-0
UN: 2810
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR