4-Chlooraniline 98%
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
108590010 108590050 108591000 108595000
ACRO108590010EA 92.7 EUR
ACRO108590010 ACRO108590050 ACRO108591000 ACRO108595000
4-Chlooraniline 98%
4-Chlooraniline
Formule: C₆H₆ClN
MW: 127.57 g/mol
Kookpunt: 232 °C (760 mmHg)
Smeltpunt: 68…72 °C
Vlampunt: >188 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00007835
CAS nummer: 106-47-8
EINECS: 203-401-0
UN: 2018
ADR: 6.1 II
Merck Index: 13,02136
Order Now
Specification Test Results
Appearance Beige to brown to purple Crystalline solid (May darken on storage)
Infrared spectrum Conforms
Melting point 68 to 72 °C
GC ≥97.5 %