4-Chlooraniline for synthesis, Sigma-Aldrich®
Leverancier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
M-Clarity™ quality level = MQ200
8.02613.0100 8.02613.0500
8.02613.0100EA 34.7 EUR
8.02613.0100 8.02613.0500
4-Chlooraniline for synthesis, Sigma-Aldrich®
4-Chlooraniline
Formule: C₆H₆ClN
MW: 127.57 g/mol
Kookpunt: 232
Smeltpunt: 69…71 °C
Dichtheid: 1.43 g/cm³ (20 °C)
Vlampunt: 120
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00007835
CAS nummer: 106-47-8
EINECS: 203-401-0
UN: 2018
ADR: 6.1 II
Merck Index: 13,02136
Order Now
Specification Test Results
Solubility 2.2 g/l (25 °C)
Melting Point 66 - 69 °C
Molar Mass 127.57 g/mol
Bulk Density 790 kg/m3
Boiling Point 232 °C (1013 hPa)
Vapor Pressure 0.07 hPa (20 °C)
Flash Point 132 °C DIN 51758
Density 1.17 g/cm3 (70 °C)
pH 6.9 (1 g/l, H2O, 20 °C)
Ignition Point 685 °C