Wash bottles, wide neck, with printed name, LaboPlast®
Leverancier: BURKLE
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
0310-3050 0310-3054 0310-3053 0310-3052 0310-3055 0310-3051
BURK0310-3050EA 9.6 EUR
BURK0310-3050 BURK0310-3054 BURK0310-3053 BURK0310-3052 BURK0310-3055 BURK0310-3051
Wash bottles, wide neck, with printed name, LaboPlast®
Flessen Spuitflessen
LDPE yellow transparent with light protection.
  • Safe filling due to wide-necked opening
  • Good chemical resistance
  • Good temperature resistance
Wash bottle with medium overprint as well as hazard and safety instructions (health statements/physical statements) based on the GHS system with substance name in German, English, French and Spanish, GHS pictograms and CAS number.