Mixing measuring cylinder, tall form, class B, borosilicate glass, MBL®
Leverancier: SciLabware
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
CRC306 CRC308 CRC312 CRC302 CRC304 CRC310 CRC314 CRC316
612-0021EA 54.7 EUR
612-0021 612-0022 612-3877 BIBBCRC302 BIBBCRC304 BIBBCRC310 BIBBCRC314 BIBBCRC316
Mixing measuring cylinder, tall form, class B, borosilicate glass, MBL®
Cilinders Maatcilinders
Borosilicate glass, class B.
  • Supplied with polyethylene stopper
  • Graduations in easy to read blue enamel
  • Robust hexagonal foot
Certificaten: DIN EN ISO 4788