Vacuum filtering flasks, Büchner, with glass hose connection
Leverancier: SciLabware
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
1170/02M 1170/08M 1170/10D 1170/12M 1170/14D 1170/04M 1170/06M
511-0734EA 40.7 EUR
511-0734 511-0771 511-0737 511-0773 511-0738 511-0735 511-0736
Vacuum filtering flasks, Büchner, with glass hose connection
Kolven Filterkolven
PYREX®, borosilicate glass.
  • For vacuum filtration
  • Heavy walls provide increased mechanical strength
  • 'Studded' base to minimise surface abrasion
Büchner filter flasks with integral glass side arm to accept 9 mm bore flexible tubing.
Certificaten: Complies with ISO 6556