Stofmaskers, voor eenmalig gebruik, FFP2/FFP3, Smart Series®
Leverancier: Moldex
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
2480 2485 2505 2495
111-0494EA 82.6 EUR
111-0494 111-0495 111-0541 111-0540
Stofmaskers, voor eenmalig gebruik, FFP2/FFP3, Smart Series®
Ademhalingsmaskers Wegwerpmaskers Stofmaskers
These masks provide protection against fine toxic dust, oil and water-based mists and fumes. Due to the DuraMesh® structure, Moldex masks are strong and durable, thus reducing consumption. The ActivForm® seal automatically adjusts to different face types and their movements. The masks have passed the optional Dolomite clogging test and therefore ensure better breathing resistance.
  • Smart Masks:
  • Additional comfort thanks to nose seal and adjustable strap with clip
  • With Ventex® valve that reduces moisture and heat, keeping the mask as cool as possible
  • Smart