Adapters met slangaansluiting, DURAN®
Leverancier: DWK Life Sciences
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
243000604 243000801 243000904
SCOT243000604EA 15.4 EUR
SCOT243000604 SCOT243000801 SCOT243000904
Adapters met slangaansluiting, DURAN®
Adapters Adapters, Slijpstukken met slangaansluitingen
DURAN®, borosilicate glass 3.3, clear.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
Adapter with standard ground joint cone and right angled tube connector.