Adapters for distilling receivers, Bredt, bent
Leverancier: Lenz Laborglas GmbH & CO.KG
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
05285104 05285101
271-2532EA 61.5 EUR
271-2532 271-2531
Adapters for distilling receivers, Bredt, bent
Onderdelen voor destillatie Destillatieadapters
DURAN®, borosilicate glass.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Bent adapters (105°), with two NS sockets, vacuum connection: GL 18.