Buckets, Round, PP
Leverancier: Bürkle
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
BURK2306-1015EA 43.5 EUR
BURK2306-1015 BURK2306-1012
Buckets, Round, PP
Emmers
Made of PP, translucent.
  • With handle
  • With spout
  • With contents scale, graduation 1000 ml
  • Sterilisable
Bucket for transporting, decanting and mixing. Safe emptying with wide spout.