3,5-Dichloorfenol 99%
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
113610100 113610500
ACRO113610100EA 94.6 EUR
ACRO113610100 ACRO113610500
3,5-Dichloorfenol 99%
3,5-Dichloorfenol
Formule: C₆H₄Cl₂O
MW: 163 g/mol
Kookpunt: 233 °C (760 mmHg)
Smeltpunt: 64…68 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00002259
CAS nummer: 591-35-5
EINECS: 209-714-9
UN: 2020
ADR: 6.1 III
Order Now
Specification Test Results
Appearance Yellow to light brown Crystalline powder or crystals or crystalline mass
Infrared spectrum Conforms
Melting point 64 to 68 °C
GC ≥98.5 %
Heavy metals (as Pb) ≤10 ppm
Insoluble matter ≤0.005 %