Filter candles, with tube
Leverancier: Lenz Laborglas GmbH & CO.KG
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
07208220 07208320 07208020 07208120 07208420 07209025 07209125 07209225 07209325 07209425
LENZ07208220EA 49.4 EUR
LENZ07208220 LENZ07208320 LENZ07208020 LENZ07208120 LENZ07208420 LENZ07209025 LENZ07209125 LENZ07209225 LENZ07209325 LENZ07209425
Filter candles, with tube
Filters Filterbuizen
These cylindrical filter plugs with tube are made of borosilicate glass 3,3.