Flessen voor opgeloste zuurstof
Leverancier: Brand
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
386038 386048
528-1303EA 28.8 EUR
528-1303 BRND386048
Flessen voor opgeloste zuurstof
Flessen Flessen voor opgeloste zuurstof
Soda-lime glass.
  • Solid, obliquely cut standard-ground glass stopper can be secured with a fastening clip
  • Each bottle is calibrated with its own stopper; hence, stoppers and bottles are not interchangeable
  • Each bottle and its stopper is marked with a unique matching identification number
For the determination of oxygen dissolved water. The measured volume is indicated to ± 0,01 ml. White labelling area.