Flessendragers
Leverancier: BURKLE
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
0361-0006 0361-0008
BURK0361-0006EA 15.1 EUR
BURK0361-0006 BURK0361-0008
Flessendragers
Dragers Flessendragers
Made of PP or HDPE.
  • Bottle transport container (PP) for up to 6 bottles, bottle transport box for up to 8 bottles
  • Bottle transport box (HDPE): suitable for food; With impermeable base suitable as a collecting basin