Containers, wijde opening, vierkant, met schroefdoppen
Leverancier: BURKLE
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
0355-0300 0357-0200 0357-1000 0355-2000 0357-0500 0357-0050 0357-0100 0355-0200 0356-0100 0356-0200 0355-1000 0355-0100 0355-0050 0355-0500 0355-0750
BURK0355-0300EA 2.4 EUR
BURK0355-0300 BURK0357-0200 BURK0357-1000 BURK0355-2000 BURK0357-0500 BURK0357-0050 BURK0357-0100 BURK0355-0200 BURK0356-0100 BURK0356-0200 BURK0355-1000 BURK0355-0100 BURK0355-0050 BURK0355-0500 BURK0355-0750
Containers, wijde opening, vierkant, met schroefdoppen
Monsternamepotjes Containers voor algemeen gebruik
Made of PETG, crystal clear, PVC, highly transparent or PVC, brown transparent.
  • Screw lid PP, black, seal made of PE
  • Good temperature resistance
  • Good chemical resistance
The wide opening of the container enables easy filling with liquid, powder or granules such as tablets, capsules, dyes, shipments of samples, specimens etc. Also suitable for gift packages and hobby articles. For light-sensitive material, brown transparent wide-necked containers are particularly suited.
Opgelet: *De product(en) op deze pagina vallen buiten het toepassingsgebied van de EU-richtlijn 2019/904