IV peripheral catheters, Insyte™
Leverancier: BD Medical
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
381344 381357 381323UK 381312UK 381223UK
BDAM381344EA 131 EUR
BDAM381344 BDAM381357 BDAM381323UK BDAM381312UK 720-2782
IV peripheral catheters, Insyte™
Katheters
Insyte IV peripheral venous catheters are made of Vialon™ – a unique proprietary biomaterial, which has been proven to enhance clinical outcomes.
  • Increases the ease of penetration
  • Less risk of infiltration (and phlebitis)
  • Exhibit greater kink resistance, with or without wings
* Vialon biomaterial has a tensile strength of 520 kg/cm², which is over 50% higher than the other materials. This bioengineered material is a member of polyurethanes, thus giving better hemocompatibility properties. In addition, the material softens in the body. This polymerMore Product Information