VITLAB® reagensflessen, PFA, smalle hals, BrandTech
Leverancier: BURKLE
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
0340-0500 0340-0250 0340-1000 0340-0100 0340-2500
BURK0340-0500EA 3.3 EUR
BURK0340-0500 BURK0340-0250 BURK0340-1000 BURK0340-0100 BURK0340-2500
VITLAB® reagensflessen, PFA, smalle hals, BrandTech
Flessen Kunststof flessen
HDPE, translucent, with tamper evident blue PP screw cap.
  • Suitable for use with fodstuffs
  • Good chemical resistance
  • Good temperature resistance
  • Cap absolutely tight closing