Flessendragers
Leverancier: KARTELL
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
395 396
KART395EA 97.7 EUR
KART395 KART396
Flessendragers
Dragers Flessendragers
PP, coloured.
  • Ergonomic design of the handle
  • Can be easily stacked
  • Very good chemical and temperature resistance
Designed to carry 6 and 4 bottles, with maximum diameter up to 90 mm and 110 mm respectively.
Order Now
Specificaties
Environmentally Preferable