2,5-Dichlorothiophenol
Leverancier: Thermo Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
BTB06657.1 BTB06657.10
MAYBBTB06657.1EA 0 EUR
MAYBBTB06657.1 MAYBBTB06657.10
2,5-Dichlorothiophenol
2,5-Dichlorothiophenol
Formule: C₆H₄Cl₂S
Kookpunt: 202 °C (1013 hPa)
Smeltpunt: 28 °C
Dichtheid: 1,415 g/cm³ (30 °C)
MDL Nummer: MFCD00004831
CAS nummer: 5858-18-4
EINECS: 227-486-9
UN: 2810
ADR: 6.1,III
Order Now
Specificaties
Environmentally Preferable