Wire baskets, Remanit® 4301
Leverancier: RSG
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
120.010 120.012 120.014 120.016 120.018 120.020 120.030 120.040
RSGA120.010EA 59.2 EUR
RSGA120.010 RSGA120.012 RSGA120.014 RSGA120.016 RSGA120.018 RSGA120.020 RSGA120.030 RSGA120.040
Wire baskets, Remanit® 4301
Manden Draadmanden
These electropolished wire baskets are made of stainless steel ¹⁸/₁₀.
  • Very good temperature resistance
  • Good chemical resistance
  • Reinforced edges