Petrischalen
Leverancier: Simport Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
D210-18B D210-18A
720-0396EA 315 EUR
720-0396 SIMPD210-18A
Petrischalen
Cultuurschalen Petrischalen
PS, sterile. These 50×9 mm Petri dishes with absorbent pads are ideal for culturing micro-organisms on either agar or broth based media.
  • Stackable
  • Squared off edges on both the top and bottom provide convenient grips for one handed opening
  • Snug fitting top ensures that neither the absorbent pad nor the agar media will dry out during incubation
  • Frosted area on top of lid permits labelling
They are designed to accommodate 47 mm diameter membrane filters.