Trays, general purpose
Leverancier: SciLabware
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
1490/04 1490/06 1490/08
216-0799EA 175 EUR
216-0799 216-0809 216-0810
Trays, general purpose
Reservoirs/Schalen/Potten Laboratoriumbakken/-schalen
PYREX®, borosilicate glass.
  • Chemically resistant
  • Can be autoclaved and used in microwave ovens
  • With handles
Ideal for numerous applications, drying powders and crystals, staining and development, collection of drips and spills from solutions bottles, gel electrophoresis, paper chromatography and dissection work.