Measuring cylinders, tall form, class A, borosilicate glass, PYREX®
Leverancier: SciLabware
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
3201/08 3201/10 3201/12 3201/14 3201/16 3201/02 3201/04 3201/06 3201/18 3201/02WC 3201/04WC 3201/06WC 3201/08WC 3201/10WC 3201/12WC 3201/14WC 3201/16WC 3201/18WC
612-3882EA 29.4 EUR
612-3882 612-3883 612-3884 612-3885 612-3886 612-3879 612-3880 612-3881 612-3887 BIBB3201/02WC BIBB3201/04WC BIBB3201/06WC BIBB3201/08WC BIBB3201/10WC BIBB3201/12WC BIBB3201/14WC BIBB3201/16WC BIBB3201/18WC
Measuring cylinders, tall form, class A, borosilicate glass, PYREX®
Cilinders Maatcilinders
Borosilicate glass, tall form, class A.
  • Hexagonal glass base, with spout
  • Easy to read white enamel graduations
ISO 4788
Certificaten: ISO 4788