Vacuum filtering flasks, Büchner, with screwthread connector
Leverancier: SciLabware
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
1170/10SC 1170/02SC 1170/14SC 1170/04SC 1170/06SC 1170/08SC
511-0777EA 164 EUR
511-0777 511-0775 511-0785 511-0045 511-0046 511-0047
Vacuum filtering flasks, Büchner, with screwthread connector
Kolven Filterkolven
PYREX®, borosilicate glass.
  • Side arm features a PTFE screwthread connector for easy and safe fitting of 9 mm bore vacuum tubing
  • With tooled neck to accept a rubber stopper or cone
Büchner flasks with heavy wall construction for vacuum filtration applications.
Certificaten: Complies with ISO 6556