Soxhlet extractors, complete assemblies
Leverancier: SciLabware
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
100RASX 200RASX 600RDSX
537-0022EA 328 EUR
537-0022 537-0023 BIBB600RDSX
Soxhlet extractors, complete assemblies
Extractors Soxhlet-extractors en Soxhlet-type extractors
PYREX®, borosilicate glass.
  • Complete glass assembly
  • Low solvent volumes required for extraction of soluble components in solids
  • Range of sizes of extractor to meet different sample sizes
* 75 mm diameter extractor flange.
Leveringsinformatie: Assembly consists of round bottom flask, Soxhlet extractor and Allihn condenser.