Graduated pipettes, type 1, Class AS, MBL
Leverancier: SciLabware
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
PRA770AS PRA772AS PRA774AS PRA776AS PRA778AS
612-3940EA 9.3 EUR
612-3940 612-3941 612-3942 612-3943 612-3944
Graduated pipettes, type 1, Class AS, MBL
Pipetten Gegradueerde pipetten
Soda lime glass.
  • Calibrated to deliver from zero at the top to any graduation line down to the shoulder
  • Graduations in permanent amber stain
DIN EN ISO 835
Certificaten: DIN EN ISO 835.