Flessen, brede hals, met schroefdoppen, PTFE
Leverancier: KARTELL
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
745 746 747 744
KART745EA 292 EUR
KART745 KART746 KART747 KART744
Flessen, brede hals, met schroefdoppen, PTFE
Flessen Kunststof flessen
PTFE, white.
  • Very good mechanical and chemical resistance
  • Can be safely used continuously from −270° C up to 250 °C
  • Suitable for foodstuff
Particularly suitable for dissolutions and digestions with acids and corrosive agents. Entire bottle, including screw caps and inserts made of PTFE.