Tissue processing/embedding cassettes in QuickLoad™, M480SL and M480T
Leverancier: Simport Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
M480-10T M480-11T M480-12T M480-2T M480-3T M480-4T M480-5T M480-6T M480-7T M480-8T M480-9T M480-10SL M480-11SL M480-12SL M480-2SL M480-3SL M480-4SL M480-5SL M480-6SL M480-7SL M480-8SL M480-9SL
720-1923EA 464 EUR
720-1923 720-1925 720-1927 720-1929 720-1931 720-1933 720-1935 720-1937 720-1939 720-1941 720-1943 720-1922 720-1924 720-1926 720-1928 720-1930 720-1932 720-1934 720-1936 720-1938 720-1940 720-1942
Tissue processing/embedding cassettes in QuickLoad™, M480SL and M480T
Histologie-cassettes Cassettes voor weefselbehandeling
Acetal polymer. Sleeved cassettes especially made to be used with Thermo Fisher printers. Cassettes stacked with tape should be used with Leica and Sakura inkjet printers.
  • The efficient flow-through slots maximise fluid exchange and ensure proper reagent drainage
  • Keep specimens safely submerged
  • Totally resistant to the chemical action of solvents used in histology laboratories