Pentafluorobenzaldehyde 98%
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
Synoniemen: 2,3,4,5,6-Pentafluorobenzaldehyde
147090025 147090100 147090500
ACRO147090025EA 34.5 EUR
ACRO147090025 ACRO147090100 ACRO147090500
Pentafluorobenzaldehyde 98%
Pentafluorobenzaldehyde
Formule: C₇HF₅O
Kookpunt: 164…166 °C (760 mmHg)
Smeltpunt: 20 °C
Dichtheid: 1,588 g/cm³
Vlampunt: 77 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00003303
CAS nummer: 653-37-2
EINECS: 211-502-6
UN: 2810
ADR: 6.1,III
Order Now
Specification Test Results
Appearance Clear yellow Liquid after melting
Infrared spectrum Conforms
Refractive index 1.4490 to 1.4510 (20°C, 589 nm)
GC ≥97.5 %