2,4-Difluorobenzonitrile
Leverancier: Thermo Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
CD07877.1 CD07877.10 CD07877.50
MAYBCD07877.1EA 0 EUR
MAYBCD07877.1 MAYBCD07877.10 MAYBCD07877.50
2,4-Difluorobenzonitrile
2,4-Difluorobenzonitrile
Formule: C₇H₃F₂N
MW: 139.1 g/mol
Kookpunt: 80…83 °C (22.5 mmHg)
Smeltpunt: 46…47 °C
MDL Nummer: MFCD00009826
CAS nummer: 3939-09-1
EINECS: 223-523-8
UN: 3439
ADR: 6.1 III