4-Fluorobenzonitrile 99%
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
Synoniemen: p-Fluorobenzonitrile
160810050 160810250
ACRO160810050EA 28.8 EUR
ACRO160810050 ACRO160810250
4-Fluorobenzonitrile 99%
4-Fluorobenzonitrile
Formule: C₇H₄FN
MW: 121.11 g/mol
Kookpunt: 188 °C (750 mmHg)
Smeltpunt: 33…36 °C
Vlampunt: 65
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00001812
CAS nummer: 1194-02-1
EINECS: 214-784-9
UN: 3439
ADR: 6.1 III
Order Now
Specification Test Results
Appearance White Low melting crystalline solid
Infrared spectrum Conforms
Melting point 33 to 36 °C
GC ≥98.5 %
Stabilizer 0.1 % 4-tert-Butylcatechol