4-Fluoro-3-nitrobenzoic acid 98%
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
305060050 305060250
ACRO305060050EA 35.2 EUR
ACRO305060050 ACRO305060250
4-Fluoro-3-nitrobenzoic acid 98%
4-Fluoro-3-nitrobenzoic acid
Formule: C₇H₄FNO₄
MW: 185.11 g/mol
Kookpunt: 92 °C (15 mmHg)
Smeltpunt: 120…126 °C
MDL Nummer: MFCD00007058
CAS nummer: 453-71-4
EINECS: 207-221-3
Order Now
Specification Test Results
Appearance Light yellow to tan Powder or crystals
Infrared spectrum Conforms
Melting point 119 to 126 °C
GC ≥97.5 %