1,4-Difluoro-2-nitrobenzene

Leverancier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

SB01118.1 SB01118.10 SB01118.50
MAYBSB01118.1EA 0 EUR
MAYBSB01118.1 MAYBSB01118.10 MAYBSB01118.50
1,4-Difluoro-2-nitrobenzene
1,4-Difluoro-2-nitrobenzene
Formule: C₆H₃F₂NO₂
Kookpunt: 207 °C (1013 hPa)
Dichtheid: 1,4671 g/cm³ (17 °C)
MDL Nummer: MFCD00007054
CAS nummer: 364-74-9
EINECS: 206-663-4
UN: 2810
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR