(Trifluoromethoxy)benzene ≥99%
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
B25658.14 B25658.22
B25658.14EA 47.5 EUR
B25658.14 B25658.22
(Trifluoromethoxy)benzene ≥99%
(Trifluoromethoxy)benzene
Purity: 99% CAS No.: 456-55-3 Molecular Formula: C7H5F3O Formula Weight: 162.11 Flash Point: 12°(53°F) Refractive Index: 1.4060 Odour: Aromatic Boiling Point: 102-104° Beilstein Reference: 2043132 UN No.: 1993 MDL No.: MFCD00040832
Formule: C₇H₅F₃O
Kookpunt: 102…104 °C
Smeltpunt: –50 °C
Dichtheid: 1,226
Vlampunt: 12 °C (53 °F)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00040832
CAS nummer: 456-55-3
EINECS: 207-269-5
UN: 1993
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=faf76f9047ed23a3eb2a61f855972f76a124b68f&vid=a016508dad7369160c1bbf2e340b9782ac2b4ec4&prid=web&clid=SAPDAM