4-Iodobenzonitrile 99%
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
Synoniemen: p-Iodobenzonitrile
412380250 412380050
ACRO412380250EA 210 EUR
ACRO412380250 ACRO412380050
4-Iodobenzonitrile 99%
4-Iodobenzonitrile
Formule: C₇H₄IN
MW: 229.02 g/mol
Kookpunt: 285
Smeltpunt: 125…128 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00051310
CAS nummer: 3058-39-7
UN: 3439
ADR: 6.1 III
Order Now
Specification Test Results
Appearance Light yellow to yellow-brown Powder and/or chunks
Infrared spectrum Conforms
Melting point 125 to 128 °C
GC ≥98.5 %
Stabilizer 50 to 150 ppm Amylene