5-Iodoanthranilic acid 98%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

A19838.06 A19838.14
A19838.06EA 13.8 EUR
A19838.06 A19838.14
5-Iodoanthranilic acid 98%
5-Iodoanthranilic acid

Purity: 98% CAS No.: 5326-47-6 Molecular Formula: C7H6INO2 Formula Weight: 263.03 Melting Point: ca 220° dec. Beilstein Reference: 639029

Formule: C₇H₆INO₂
Kookpunt: 360 °C (1013 hPa)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00007849
CAS nummer: 5326-47-6
EINECS: 226-205-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR