Slangen van PTFE
Leverancier: KARTELL
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
3901 3902 3903 3904 3905 3906 3908 3907 3900
KART3901EA 113 EUR
KART3901 KART3902 KART3903 KART3904 KART3905 KART3906 KART3908 KART3907 KART3900
Slangen van PTFE
Slangen Transfer Tubing
PTFE, white.
  • Excellent chemical resistance
  • Very good thermal resistance, autoclavable
  • Free of extractable substances, physiologically neutral, biocompatible
Excellent resistance to acids, bases and solvents.
Temperature resistance: −200 to +260 °C