Tissue cassettes in QuickLoad™ taped stacks, Unisette™, M505T
Leverancier: Simport Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
M505-10T M505-11T M505-12T M505-2T M505-3T M505-4T M505-5T M505-6T M505-7T M505-8T M505-9T
720-1389EA 213 EUR
720-1389 720-1390 720-1391 720-1392 720-1393 720-1394 720-1395 720-1396 720-1397 720-1398 720-1399
Tissue cassettes in QuickLoad™ taped stacks, Unisette™, M505T
Histologie-cassettes Cassettes voor weefselbehandeling
Moulded from a special high density acetal polymer, these cassettes keep specimens safely submerged in liquid and are totally resistant to the chemical action of histological solvents.
  • M506 series is similar to M505 but specially designed to hold biopsy specimens during the embedding process
  • Available in non cytotoxic, non metallic colours
  • The snap-latch and hinge-lock design prevents early separation of base and lid and allows for one handed operation
The efficient flow-through slots maximise fluid exchange and ensure proper drainage. The one-piece integral lid eliminates the need for separate steel lids. Lids can be opened and closed as often as necessary and they always relock securely without danger of specimen loss. Large labelling areas are located on two sides of the cassettes. Anterior writing area is at a 35° angle.
Verpakking: 40×25 taped stacks for a total of 1000 cassettes.