Biopsy cassettes in QuickLoad™ taped stacks, Unisette™, M506T
Leverancier: Simport Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
M506-10T M506-11T M506-12T M506-2T M506-3T M506-4T M506-5T M506-6T M506-7T M506-8T M506-9T
720-1400EA 241 EUR
720-1400 720-1401 720-1402 720-1403 720-1404 720-1405 720-1406 720-1407 720-1408 720-1409 720-1410
Biopsy cassettes in QuickLoad™ taped stacks, Unisette™, M506T
Histologie-cassettes Cassettes voor biopsiebehandeling
Moulded from a special high density acetal polymer, these cassettes keep specimens safely submerged in liquid and are totally resistant to the chemical action of histological solvents.
  • The snap-latch and hinge-lock design prevents early separation of base and lid and allows for one handed operation
  • Large labelling areas are located on two sides of the cassettes
  • Available in several colours
The efficient flow-through slots maximise fluid exchange and ensure proper drainage. The one-piece integral lid eliminates the need for separate steel lids. Lids can be opened and closed as often as necessary and they always relock securely without danger of specimen loss. Anterior writing area is at a 35° angle. M506 series is similar to M505 but specially designed to hold biopsy specimens during the embedding process.
Verpakking: 40×25 taped stacks for a total of 1000 cassettes.