Flessen, brede hals, rond, met schroefdop, WHEATON®
Leverancier: DWK Life Sciences
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
209673 209674 209675 209676
WHEA209673EA 266 EUR
WHEA209673 WHEA209674 WHEA209675 WHEA209676
Flessen, brede hals, rond, met schroefdop, WHEATON®
Flessen Kunststof flessen
HDPE, natural, with with PP screw cap.
  • Good chemical resistance
  • Pre-attached screw cap with foamed PE liner
  • Good thermal resistance
Wide mouth ideal for solids.