Slangen, rubber
Leverancier: KARTELL
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
3860 3861 3862 3863 3864 3865 3866 3867 3868
KART3860EA 65.9 EUR
KART3860 KART3861 KART3862 KART3863 KART3864 KART3865 KART3866 KART3867 KART3868
Slangen, rubber
Slangen Transfer Tubing
Natural rubber (NR), orange.
  • Temperature resistance: −35 to +70 °C
Useful for gases, non-corrosive liquids and medium concentration acids.