Stainless steel, gauze woven from wire for gas burners
Leverancier: RSG
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
247.875 247.855 247.835 247.815 247.895
RSGA247.875EA 45.6 EUR
RSGA247.875 RSGA247.855 RSGA247.835 RSGA247.815 RSGA247.895
Stainless steel, gauze woven from wire for gas burners
Stainless steel
Wire mesh out of stainless steel wire with folded-up borders
  • Stainless steel 1.4301
  • with folded up edge
  • mesh: 0.6 x 2.0 mm
Order Now
Specificaties
Environmentally Preferable