Watermonsternemers, AquaSampler
Leverancier: BURKLE
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
5308-0350 5308-1050 5308-0700
300-0078EA 395 EUR
300-0078 300-0080 300-0079
Watermonsternemers, AquaSampler
Monsternemers Monsternemers voor water
Samplers available in three sizes.
  • Ultra pure materials guaranteeing high purity
  • Easy to clean with rounded edges and threads
  • FEP/PTFE ultra pure and anti-adhesive surface
Designed for ground water samples from wells, pits, and other sources.