Glucose and lactate determination tubes, BD Vacutainer®
Leverancier: BD Medical
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
368920UK 368920 368521 368521UK
BDAM368920UKEA 193 EUR
BDAM368920UK BDAM368920 BDAM368521 BDAM368521UK
Glucose and lactate determination tubes, BD Vacutainer®
Diagnostische systemen Clinical Sample Collection/Preparation Blood Sample Collection/Preparation
Glass, clear.
  • With label
  • Hemogard™ closure
These tubes used for the determination of blood sugar are available with two different additives fluoride and iodoacetate.