Aniline 99+% ACS
Leverancier: Thermo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
36238.K2EA
153
EUR
ABCRAB207948.0100
36238.K2
36238.AE
36238.AP
Aniline 99+% ACS
Aniline
Purity: 99+%
CAS No.: 62-53-3
Molecular Formula: C6H7N
Formula Weight: 93.13
Flash Point: 70°(158°F)
Physical Form: Liquid
Density: 1.021
Refractive Index: 1.5860
Melting Point: -6°
Boiling Point: 183-184°
Beilstein Reference: 605631
Merck Reference: 14,659
UN No.: 1547
MDL No.: MFCD00007629
Formule:
C₆H₅NH₂ MW: 93,13 g/mol Kookpunt: 183…184 °C Smeltpunt: –6 °C Dichtheid: 1,021 Vlampunt: 70 °C (158 °F) Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00007629 CAS nummer: 62-53-3 EINECS: 200-539-3 UN: 1547 ADR: 6.1,II Merck Index: 13,00661 |