Glass cutter knife
Leverancier: RSG
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
267.100 267.101
RSGA267.100EA 101 EUR
RSGA267.100 RSGA267.101
Glass cutter knife
Snijders
Plastic handle with hardened metal blade.
  • Clean and precise cutting
  • Replaceable hardened metal blades
  • Easy to change blade
Blade size 40x20x1,5 mm.