Dispenser bottles, Wheaton®
Leverancier: DWK Life Sciences
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
211195 211194
WHEA211195EA 321 EUR
WHEA211195 WHEA211194
Dispenser bottles, Wheaton®
Flessen Druppelflessen
These translucent dispensing bottles are excellent for mild and strong buffers with a spout to dispense the contents and then could be depressed to seal the bottle.
  • Very flexible with high impact strength
  • Good chemical resistance