Bottles, narrow neck, borosilicate glass, clear or amber, with standard ground joints, WHEATON®
Leverancier: DWK Life Sciences
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
215237 215239 215240 215241 215243 215257 215259 W215235 W215255
WHEA215237EA 599 EUR
WHEA215237 WHEA215239 WHEA215240 WHEA215241 WHEA215243 WHEA215257 WHEA215259 WHEAW215235 WHEAW215255
Bottles, narrow neck, borosilicate glass, clear or amber, with standard ground joints, WHEATON®
Flessen Glazen flessen
Bottles, made of borosilicate glass, with glass stopper.
  • Very good chemical and temperature resistance
  • Amber coloured for light sensitive substances
  • Octagon-shaped round glass stopper fits securely to prevent contamination