Stampers, getand, PTFE-kop
Leverancier: KARTELL
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
6315 6302 6305 6310 6330 6350
KART6315EA 88.7 EUR
KART6315 KART6302 KART6305 KART6310 KART6330 KART6350
Stampers, getand, PTFE-kop
Vijzels en stampers Stampers
Pestles used in the combination with borosilicate mortar for sample crushing. Ideal for tough materials such as tumours.
  • Head made of PTFE
  • Shaft made of stainless steel